Supelco® PESTANAL® Organic Reference Standard, Simazine-D₁₀
34054-10MG-R
: Merck
:
- Pk:10 mg
- Standards:PESTANAL®, Analytical standard
- CAS No.:220621-39-6
- Description:Simazine-d₁₀
- Storage conditions:2...30 °C
- Hazard class description:H351 - H410
- Shipping conditions:STD
- MDL No.:MFCD01632307
- M-Clarity quality level:MQ100
- Synonym(s):2,4-Bis(pentadeuteroethylamino)-6-chloro-1,3,5-triazine
- Formula:C₇H₂D₁₀ClN₅
- Molecular weight:211,72
- Flash point:Not applicable
- GHS signal: GHS08, GHS09
: P202 - P273 - P280 - P308 + P313 - P391 - P405