Order Entry
Northern Ireland
ContactUsLinkComponent
Supelco® PESTANAL® Organic Reference Standard, Pyrimidifen
Supelco® PESTANAL® Organic Reference Standard, Pyrimidifen
Catalog # 35999-10MG
Supplier:  Merck
CAS Number:  
undefined
Supelco® PESTANAL® Organic Reference Standard, Pyrimidifen
Catalog # 35999-10MG
Supplier:  Merck
CAS Number:  

Specifications

  • Pk:
    10 mg
  • Standards:
    PESTANAL®, Analytical standard
  • CAS No.:
    105779-78-0
  • Description:
    Pyrimidifen
  • Storage conditions:
    2...8 °C
  • Hazard class description:
    H301 - H410
  • Shipping conditions:
    WICE
  • MDL No.:
    MFCD01631162
  • M-Clarity quality level:
    MQ100
  • Synonym(s):
    5-Chloro-N-{2-[4-(2-ethoxyethyl)-2,3-dimethylphenoxy]ethyl}-6-ethylpyrimidin-4-amine
  • Formula:
    C₂₀H₂₈ClN₃O₂
  • Molecular weight:
    377,91
  • Flash point:
    Not applicable

Specifications

About this item

  • GHS signal: GHS06, GHS09
Caution: P264 - P273 - P301 + P310 - P391 - P405 - P501