Organic reference standard, Propyl 4-hydroxybenzoate-ring-13C6 solution, analytical standard, Supelco®
Catalog # 32562-1ML
Supplier: Merck
CAS Number:
Specifications
- Concentration:50 µg/ml in acetone
- Pk:1 mL
- Standards:Analytical standard
- Description:Propyl 4-hydroxybenzoate-ring-13C6 solution
- Formula:13C₆C₄H₁₂O₃
- Storage temperature:−20 °C
- Description:Propyl 4-hydroxybenzoate-ring-13C6 solution