Supelco® PESTANAL® Organic Reference Standard, Prometryn
Catalog # 45636-50MG
Supplier: Merck
CAS Number:
Specifications
- Pk:50 mg
- Standards:PESTANAL®, Analytical standard
- CAS No.:7287-19-6
- Description:Prometryn
- Storage conditions:2...30 °C
- Hazard class description:H302 + H332 - H410
- Shipping conditions:STD
- MDL No.:MFCD00055396
- M-Clarity quality level:MQ100
- Synonym(s):2,4-Bis-(isopropylamino)-6-methylthio-1,3,5-triazine
- Formula:C₁₀H₁₉N₅S
- Molecular weight:241,36
Specifications
About this item
- GHS signal: GHS07, GHS09
Caution: P261 - P264 - P271 - P273 - P301 + P312 - P304 + P340 + P312