Order Entry
Northern Ireland
ContactUsLinkComponent
 
Supelco® PESTANAL® Organic Reference Standard, Profenofos
  45632-250MG
 :  Merck
 :  
undefined
Supelco® PESTANAL® Organic Reference Standard, Profenofos
  45632-250MG
 :  Merck
 :  

 

  • Pk:
    250 mg
  • Standards:
    PESTANAL®, Analytical standard
  • CAS No.:
    41198-08-7
  • Description:
    Profenofos
  • Storage conditions:
    2...8 °C
  • Hazard class description:
    H302 + H312 + H332 - H317 - H410
  • Shipping conditions:
    STD
  • MDL No.:
    MFCD00078736
  • M-Clarity quality level:
    MQ100
  • Formula:
    C₁₁H₁₅BrClO₃PS
  • Molecular weight:
    373,63
  • Storage temperature:
    2...8 °C
  • Flash point:
    124 °C - closed cup

 

 

  • GHS signal: GHS07, GHS09
 : P261 - P273 - P280 - P301 + P312 - P302 + P352 + P312 - P304 + P340 + P312