Order Entry
Northern Ireland
ContactUsLinkComponent
 
Supelco® PESTANAL® Organic Reference Standard, Pinoxaden
Catalog # 32821-25MG
Supplier:  Merck
CAS Number:  
undefined
Supelco® PESTANAL® Organic Reference Standard, Pinoxaden
Catalog # 32821-25MG
Supplier:  Merck
CAS Number:  

Specifications

  • Pk:
    25 mg
  • Standards:
    PESTANAL®, Analytical standard
  • CAS No.:
    243973-20-8
  • Description:
    Pinoxaden
  • Storage conditions:
    2...30 °C
  • Hazard class description:
    H302 + H332 - H317 - H319 - H335 - H361d - H410
  • Shipping conditions:
    STD
  • MDL No.:
    MFCD08690563
  • M-Clarity quality level:
    MQ100
  • Formula:
    C₂₃H₃₂N₂O₄
  • Molecular weight:
    400,51
  • Flash point:
    Not applicable

Specifications

About this item

  • GHS signal: GHS07
Caution: P273 - P280 - P301 + P312 - P302 + P352 - P304 + P340 + P312 - P308 + P313