Supelco® PESTANAL® Organic Reference Standard, Picloram
Catalog # 36774-250MG-R
Supplier: Merck
CAS Number:
Specifications
- Pk:250 mg
- Standards:PESTANAL®, Analytical standard
- CAS No.:1918-02-1
- Description:Picloram
- Storage conditions:2...30 °C
- Shipping conditions:STD
- MDL No.:MFCD00012101
- M-Clarity quality level:MQ100
- Synonym(s):4-Amino-3,5,6-trichloropyridine-2-carboxylic acid; 4-Amino-3,5,6-trichloropicolinic acid; Picloram
- Melting point:200 °C (dec.) (lit.)
- Formula:C₆H₃Cl₃N₂O₂
- Molecular weight:241,46
- Flash point:Not applicable
Specifications
About this item
- GHS signal: GHS07