Organic reference standard, Padimate O, analytical standard, Supelco®
Catalog # 74309-1ML
Supplier: Merck
CAS Number:
Specifications
- Pk:1 mL
- Standards:Analytical standard
- CAS No.:21245-02-3
- Description:Padimate O
- Formula:C₁₇H₂₇NO₂
- Description:Padimate O
- Boiling point:325 °C (lit.)
Specifications
About this item
Padimate O is a tertiary amine derivative of p-aminobenzoic acid (PABA), which is commonly used as a sunscreen agent in cosmetics and other sunscreen formulations due to its ability to absorb ultraviolet radiations.
Padimate O may be used as a reference standard in the determination of padimate O in cosmetics using high performance liquid chromatography (HPLC).