Order Entry
Northern Ireland
ContactUsLinkComponent
Organic reference standard, Nonanal 2,4-dinitrophenylhydrazone, analytical standard, Supelco®
Organic reference standard, Nonanal 2,4-dinitrophenylhydrazone, analytical standard, Supelco®
Catalog # 33851-100MG
Supplier:  Merck
CAS Number:  
undefined
Organic reference standard, Nonanal 2,4-dinitrophenylhydrazone, analytical standard, Supelco®
Catalog # 33851-100MG
Supplier:  Merck
CAS Number:  

Specifications

  • Pk:
    100 mg
  • Standards:
    Analytical standard
  • CAS No.:
    2348-19-8
  • Description:
    Nonanal 2,4-dinitrophenylhydrazone
  • M-Clarity quality level:
    MQ100
  • Melting point:
    99...103 °C
  • Formula:
    C₁₅H₂₂N₄O₄
  • Description:
    Nonanal 2,4-dinitrophenylhydrazone

Specifications

About this item

Packaging: Bottomless glass bottle contents are inside inserted fused cone.