Order Entry
Northern Ireland
ContactUsLinkComponent
Organic reference standard, N-Nitrosodi-n-propylamine solution, certified reference material, Supelco®
Organic reference standard, N-Nitrosodi-n-propylamine solution, certified reference material, Supelco®
Catalog # SUPL40061
Supplier:  Merck
CAS Number:  
Organic reference standard, N-Nitrosodi-n-propylamine solution, certified reference material, Supelco®
Catalog # SUPL40061
Supplier:  Merck
CAS Number:  

Specifications

  • Concentration:
    5000 µg/ml in methanol
  • Pk:
    1
  • Standards:
    Certified reference material
  • CAS No.:
    621-64-7
  • Description:
    N-Nitrosodi-n-propylamine solution
  • Formula:
    C₆H₁₄N₂O
  • Storage temperature:
    2…8 °C
  • Description:
    N-Nitrosodi-n-propylamine solution
  • Pk Info:
    1 ml/ampule

Specifications

About this item

Certifications: This certified reference material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025:2005.