Order Entry
Northern Ireland
ContactUsLinkComponent
Organic reference standard, N,N'-(Methylenediphenylene)bis{4-(2-pyridinyl)-1-piperazinecarboxamide} (derivative of 4,4'-MDIP), certified reference material, Supelco®
Organic reference standard, N,N'-(Methylenediphenylene)bis{4-(2-pyridinyl)-1-piperazinecarboxamide} (derivative of 4,4'-MDIP), certified reference material, Supelco®
Catalog # 48147.
Supplier:  Merck
CAS Number:  
Organic reference standard, N,N'-(Methylenediphenylene)bis{4-(2-pyridinyl)-1-piperazinecarboxamide} (derivative of 4,4'-MDIP), certified reference material, Supelco®
Catalog # 48147.
Supplier:  Merck
CAS Number:  

Specifications

  • Concentration:
    1000 μg/ml in DMSO (as the derivative)
  • Pk:
    1
  • Standards:
    Certified reference material
  • Description:
    N,N'-(Methylenediphenylene)bis{4-(2-pyridinyl)-1-piperazinecarboxamide} (derivative of 4,4'-MDIP)
  • Storage temperature:
    RT
  • Description:
    N,N'-(Methylenediphenylene)bis{4-(2-pyridinyl)-1-piperazinecarboxamide} (derivative of 4,4'-MDIP)
  • Pk Info:
    1 ml/ampule

Specifications

About this item

Certifications: This certified reference material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025:2005.