Supelco® TraceCERT® Organic Reference Standard, Methyl tert-butyl ether solution
Methyl tert-butyl ether solution, Certified reference material, TraceCERT®, 1 mL
Catalog # CRM48483
Supplier: Merck
Supelco® TraceCERT® Organic Reference Standard, Methyl tert-butyl ether solution
Catalog # CRM48483
Supplier: Merck
CAS Number:
Jump to:
Product Details & Documents
- GHS signal: GHS02, GHS06, GHS08
Caution: P210 - P280 - P301 + P310 + P330 - P302 + P352 + P312 - P304 + P340 + P311
Specifications
Specifications
- Pk:1 mL
- Standards:Certified reference material, TraceCERT®
- Description:Methyl tert-butyl ether solution
- Cat. No.:CRM48483
- M-Clarity quality level:MQ100
- Storage conditions:2...30 °C
- Hazard class description:H225 - H301 + H311 + H331 - H370
- Flash point:9,7 °C - Closed cup
- Shipping conditions:STD
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



