Supelco® Organic Reference Standard, Methyl Cis-11-Octadecenoate
Catalog # 17264-100MG
Supplier: Merck
CAS Number:
Specifications
- Pk:100 mg
- Standards:Analytical standard
- CAS No.:1937-63-9
- Description:Methyl cis-11-octadecenoate
- Storage conditions:–20 °C
- Hazard class description:H315 - H319 - H335
- Shipping conditions:WICE
- MDL No.:MFCD00010457
- M-Clarity quality level:MQ100
- Formula:C₁₉H₃₆O₂
- Molecular weight:296,49
- Storage temperature:–20 °C
- Flash point:62,0 °C - closed cup
Specifications
About this item
- GHS signal: GHS07
Caution: P261 - P264 - P271 - P280 - P302 + P352 - P305 + P351 + P338