Order Entry
Northern Ireland
ContactUsLinkComponent
Organic reference standard, Methyl 4-hydroxybenzoate-ring-13C6 solution, analytical standard, Supelco®
Organic reference standard, Methyl 4-hydroxybenzoate-ring-13C6 solution, analytical standard, Supelco®
Catalog # 32557-1ML
Supplier:  Merck
CAS Number:  
undefined
Organic reference standard, Methyl 4-hydroxybenzoate-ring-13C6 solution, analytical standard, Supelco®
Catalog # 32557-1ML
Supplier:  Merck
CAS Number:  

Specifications

  • Concentration:
    50 µg/ml in acetone
  • Pk:
    1 mL
  • Standards:
    Analytical standard
  • CAS No.:
    1581694-95-2
  • Description:
    Methyl 4-hydroxybenzoate-ring-13C6 solution
  • Seq:
    02
  • Formula:
    13C₆C₂H₈O₃
  • Storage temperature:
    –20 °C
  • Description:
    Methyl 4-hydroxybenzoate-ring-13C6 solution

Specifications

About this item

  • Certan vial