Pesticide reference standard, Methabenzthiazuron, PESTANAL®, analytical standard, Supelco®
36156-100MG
: Merck
:
- Pk:100 mg
- Standards:Analytical standard
- CAS No.:2032-65-7
- Description:Methabenzthiazuron
- M-Clarity quality level:MQ100
- Formula:C₁₀H₁₁N₃OS
- Description:Methabenzthiazuron