Supelco® PESTANAL® Organic Reference Standard, Metalaxyl-M
32808-100MG
: Merck
:
- Pk:100 mg
- Standards:PESTANAL®, Analytical standard
- CAS No.:70630-17-0
- Description:Metalaxyl-M
- Storage conditions:2...30 °C
- Hazard class description:H302 - H318
- Shipping conditions:STD
- MDL No.:MFCD03701742
- M-Clarity quality level:MQ100
- Synonym(s):N-(2,6-Dimethylphenyl)-N-(methoxyacetyl)-ᴅ-alanine methyl ester; Mefenoxam
- Formula:C₁₅H₂₁NO₄
- Molecular weight:279,33
- Flash point:Not applicable
- GHS signal: GHS05, GHS07
: P264 - P270 - P280 - P301 + P312 - P305 + P351 + P338 - P501