Supelco® PESTANAL® Organic Reference Standard, Isoxaflutole
Catalog # 46437-100MG
Supplier: Merck
CAS Number:
Specifications
- Pk:100 mg
- Standards:PESTANAL®, analytical standard
- CAS No.:141112-29-0
- Description:Isoxaflutole
- Storage conditions:2...30 °C
- Hazard class description:H361d - H410
- Shipping conditions:STD
- MDL No.:MFCD03095707
- M-Clarity quality level:MQ100
- Synonym(s):5-Cyclopropyl-4-[2-(methylsulfonyl)-4-(trifluoromethyl)benzoyl]isoxazole
- Molecular weight:359,32
Specifications
About this item
- GHS signal: GHS08, GHS09
Caution: P202 - P273 - P280 - P308 + P313 - P391 - P405