Order Entry
Northern Ireland
ContactUsLinkComponent
Supelco® Organic Reference Standard, Isopropyl 4-Hydroxybenzoate
Supelco® Organic Reference Standard, Isopropyl 4-Hydroxybenzoate
Catalog # 05828-25MG
Supplier:  Merck
CAS Number:  
undefined
Supelco® Organic Reference Standard, Isopropyl 4-Hydroxybenzoate
Catalog # 05828-25MG
Supplier:  Merck
CAS Number:  

Specifications

  • Pk:
    25 mg
  • Standards:
    Analytical standard
  • CAS No.:
    4191-73-5
  • Description:
    Isopropyl 4-hydroxybenzoate
  • Storage conditions:
    2...8 °C
  • Hazard class description:
    H302 - H318
  • Shipping conditions:
    STD
  • MDL No.:
    MFCD00016468
  • M-Clarity quality level:
    MQ100
  • Synonym(s):
    4-Hydroxybenzoic acid isopropyl ester; Isopropyl paraben
  • Formula:
    C₁₀H₁₂O₃
  • Molecular weight:
    180,20
  • Storage temperature:
    2...8 °C
  • Flash point:
    Not applicable

Specifications

About this item

  • GHS signal: GHS05, GHS07
Caution: P264 - P270 - P280 - P301 + P312 - P305 + P351 + P338 - P501