Supelco® PESTANAL® Organic Reference Standard, Iprovalicarb
Catalog # 33431-100MG-R
Supplier: Merck
CAS Number:
Specifications
- Pk:100 mg
- Standards:PESTANAL®, Analytical standard, mixture of isomers
- CAS No.:140923-17-7
- Description:Iprovalicarb
- Storage conditions:2...30 °C
- Hazard class description:H351
- Shipping conditions:STD
- MDL No.:MFCD04112614
- M-Clarity quality level:MQ100
- Synonym(s):Nα-(Isopropoxycarbonyl)-N-[1-(p-tolyl)ethyl]-ʟ-valinamide; Isopropyl (S)-2-methyl-1-[1-(p-tolyl)ethylcarbamoyl]propylcarbamate
- Formula:C₁₈H₂₈N₂O₃
- Molecular weight:320,43
- Flash point:Not applicable
Specifications
About this item
Caution: P201 - P202 - P280 - P308 + P313 - P405 - P501