Supelco® PESTANAL® Organic Reference Standard, Iprodione
36132-100MG
: Merck
:
- Pk:100 mg
- Standards:PESTANAL®, Analytical standard
- CAS No.:36734-19-7
- Description:Iprodione
- Storage conditions:2...30 °C
- Hazard class description:H351 - H410
- Shipping conditions:STD
- MDL No.:MFCD00055353
- M-Clarity quality level:MQ100
- Synonym(s):[3-(3,5-Dichlorophenyl)-2,4-dioxoimidazolidinyl]-N-(methylethyl)carboxamide; Iprodione
- Melting point:130...134 °C (lit.)
- Formula:Cl₂C₆H₃N₂C₃H₂O₂CONHCH(CH₃)₂
- Molecular weight:330,17
- Flash point:Not applicable
- GHS signal: GHS08, GHS09
: P202 - P273 - P280 - P308 + P313 - P391 - P405