Order Entry
Northern Ireland
ContactUsLinkComponent
Pesticide reference standard, Fusarenon X solution, analytical standard, Supelco®
Pesticide reference standard, Fusarenon X solution, analytical standard, Supelco®
  34130-2ML
 :  Merck
 :  
undefined
Pesticide reference standard, Fusarenon X solution, analytical standard, Supelco®
  34130-2ML
 :  Merck
 :  

 

  • Concentration:
    100 µg/ml in acetonitrile
  • Pk:
    2 mL
  • Standards:
    Analytical standard
  • CAS No.:
    23255-69-8
  • Description:
    Fusarenon X solution
  • M-Clarity quality level:
    MQ100
  • Formula:
    C₁₂H₈Cl₂O₃S
  • Description:
    Fusarenon X solution

 

 

Produced by Fusarium nivale fungi naturally occurring in crops such as wheat and barley.

  • Fusarenon X is a trichothecene mycotoxin