Supelco® TraceCERT® Organic Reference Standard, Formaldehyde-DNPH solution
Catalog # CRM47177
Supplier: Merck
CAS Number:
Specifications
- Concentration:100 μg/ml in acetonitrile (as the aldehyde), ampule of 1 ml
- Pk:1 mL
- Standards:Certified reference material, TraceCERT®
- CAS No.:1081-15-8
- Description:Formaldehyde-DNPH solution
- Storage conditions:2…8 °C
- Hazard class description:H225 - H302 + H312 + H332 - H319
- Shipping conditions:WICE
- MDL No.:MFCD00191364
- M-Clarity quality level:MQ100
- Synonym(s):Formaldehyde-2,4-DNPH Solution
- Formula:H₂C=NNHC₆H₃(NO₂)₂
- Molecular weight:210,15
- Flash point:2,0 °C - Closed cup
Specifications
About this item
- GHS signal: GHS02, GHS07
Certifications: ISO 34:2009 and ISO/IEC 17025:2005
Caution: P210 - P280 - P301 + P312 - P303 + P361 + P353 - P304 + P340 + P312 - P305 + P351 + P338