Pesticide reference standard, Fensulfothion PO-sulfone, PESTANAL®, analytical standard, Supelco®
Catalog # 33884-50MG-R
Supplier: Merck
CAS Number:
Specifications
- Pk:50 mg
- Standards:Analytical standard
- CAS No.:6132-17-8
- Description:Fensulfothion PO-sulfone
- Formula:C₁₁H₁₇O₆PS
- Description:Fensulfothion PO-sulfone