Supelco® PESTANAL® Organic Reference Standard, Carbophenothion
Catalog # 31461-250MG
Supplier: Merck
CAS Number:
Specifications
- Pk:250 mg
- Standards:PESTANAL®, Analytical standard
- CAS No.:786-19-6
- Description:Carbophenothion
- Storage conditions:2...8 °C
- Hazard class description:H300 - H311 - H410
- Shipping conditions:STD
- MDL No.:MFCD00128009
- M-Clarity quality level:MQ100
- Synonym(s):S-(4-Chlorophenylthiomethyl) O,O-diethyl phosphorodithioate
- Formula:C₁₁H₁₆ClO₂PS₃
- Molecular weight:342,87
- Storage temperature:2...8°C
- Flash point:Not applicable
Specifications
About this item
- GHS signal: GHS06, GHS09
Caution: P264 - P270 - P273 - P280 - P301 + P310 - P302 + P352 + P312