Supelco® Organic Reference Standard, BDE No 207 Solution
Catalog # 33617-1ML
Supplier: Merck
CAS Number:
Specifications
- Pk:1 mL
- Standards:Analytical standard
- CAS No.:437701-79-6
- Description:BDE No 207 solution
- Storage conditions:2...8 °C
- Hazard class description:H225 - H304 - H315 - H336 - H410
- Shipping conditions:STD
- MDL No.:MFCD08460509
- M-Clarity quality level:MQ100
- Synonym(s):2,2′,3,3′,4,4′,5,6,6′-Nonabromodiphenyl ether solution; 2,2′,3,3′,4,4′,5,6,6′-NonaBDE; PBDE 207
- Formula:C₁₂HBr₉O
- Molecular weight:880,27
- Flash point:–12 °C - closed cup
Specifications
About this item
- GHS signal: GHS02, GHS08, GHS07, GHS09
Caution: P210 - P273 - P301 + P310 + P331 - P302 + P352