Supelco® Organic Reference Standard, BDE No 183 Solution
33686-1ML
: Merck
:
- Pk:1 mL
- Standards:Analytical standard
- CAS No.:207122-16-5
- Description:BDE No 183 solution
- Storage conditions:2...8 °C
- Hazard class description:H225 - H304 - H315 - H336 - H410
- Shipping conditions:STD
- MDL No.:MFCD16661229
- M-Clarity quality level:MQ100
- Synonym(s):2,2′,3,4,4′,5′,6-Heptabromodiphenyl ether solution; 2,2′,3,4,4′,5′,6-HeptaBDE; PBDE 183
- Formula:C₁₂H₃Br₇O
- Molecular weight:722,48
- Storage temperature:2...8 °C
- Flash point:–12 °C
- GHS signal: GHS02, GHS08, GHS07, GHS09
: P210 - P273 - P301 + P310 + P331 - P302 + P352