Supelco® Organic Reference Standard, BDE No 100 Solution
Catalog # 33681-1ML
Supplier: Merck
CAS Number:
Specifications
- Pk:1 mL
- Standards:Analytical standard
- CAS No.:189084-64-8
- Description:BDE No 100 solution
- Storage conditions:2...8 °C
- Hazard class description:H225 - H304 - H315 - H336 - H410
- Shipping conditions:STD
- MDL No.:MFCD09037599
- M-Clarity quality level:MQ100
- Synonym(s):2,2′,4,4′,6-Pentabromodiphenyl ether solution; 2,2′,4,4′,6-PentaBDE; PBDE 100
- Formula:C₁₂H₅Br₅O
- Molecular weight:564,69
- Storage temperature:2...8 °C
- Flash point:–12 °C
Specifications
About this item
- GHS signal: GHS02, GHS08, GHS07, GHS09
Caution: P210 - P273 - P301 + P310 + P331 - P302 + P352