Order Entry
Northern Ireland
ContactUsLinkComponent
Supelco® PESTANAL® Organic Reference Standard, Amicarbazone
Supelco® PESTANAL® Organic Reference Standard, Amicarbazone
Catalog # 32396-25MG
Supplier:  Merck
CAS Number:  
undefined
Supelco® PESTANAL® Organic Reference Standard, Amicarbazone
Catalog # 32396-25MG
Supplier:  Merck
CAS Number:  

Specifications

  • Pk:
    25 mg
  • Standards:
    PESTANAL®, Analytical standard
  • CAS No.:
    129909-90-6
  • Description:
    Amicarbazone
  • Storage conditions:
    2...30 °C
  • Hazard class description:
    H302 + H332
  • Shipping conditions:
    STD
  • MDL No.:
    MFCD07364053
  • M-Clarity quality level:
    MQ100
  • Synonym(s):
    4-Amino-N-tert-butyl-4,5-dihydro-3-isopropyl-5-oxo-1,2,4-1H-triazole-1-carboxamide
  • Formula:
    C₁₀H₁₉N₅O₂
  • Molecular weight:
    241,29
  • Flash point:
    Not applicable

Specifications

About this item

  • GHS signal: GHS07
Caution: P261 - P264 - P270 - P271 - P301 + P312 - P304 + P340 + P312