Supelco® Organic Reference Standard, 4-Tert-Octylphenol-3,5-D₂ Solution
32273-1ML
: Merck
:
- Pk:1 mL
- Standards:Analytical standard
- CAS No.:1173021-20-9
- Description:4-tert-Octylphenol-3,5-d₂ solution
- Seq:01
- Storage conditions:–20 °C
- Hazard class description:H225 - H319 - H336
- Shipping conditions:WICE
- MDL No.:MFCD09842826
- M-Clarity quality level:MQ100
- Synonym(s):4-(1,1,3,3-Tetramethylbutyl)phenol-3,5-d₂; 4-tert-OP-D2
- Formula:C₁₄D₂H₂₀O
- Molecular weight:208,34
- Storage temperature:−20 °C
- Flash point:–17,0 °C - closed cup
- GHS signal: GHS02, GHS07
: P210 - P280 - P304 + P340 + P312 - P305 + P351 + P338 - P337 + P313 - P403 + P235