Order Entry
Northern Ireland
ContactUsLinkComponent
Supelco® PESTANAL® Organic Reference Standard, 3-Methylcholanthrene solution
Supelco® PESTANAL® Organic Reference Standard, 3-Methylcholanthrene solution
  46434-10ML-R
 :  Merck
 :  
undefined
Supelco® PESTANAL® Organic Reference Standard, 3-Methylcholanthrene solution
  46434-10ML-R
 :  Merck
 :  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

 

  • Concentration:
    100 μg/ml in acetonitrile
  • Pk:
    10 mL
  • Standards:
    PESTANAL®, analytical standard
  • CAS No.:
    56-49-5
  • Description:
    3-Methylcholanthrene solution
  • Seq:
    02
  • Storage conditions:
    2…8 °C
  • Hazard class description:
    H225 - H302 + H312 + H332 - H319
  • Shipping conditions:
    STD
  • MDL No.:
    MFCD00003704
  • M-Clarity quality level:
    MQ100
  • Molecular weight:
    268,35
  • Flash point:
    2,0 °C - Closed cup

 

 

  • GHS signal: GHS02, GHS07
 : P210 - P280 - P301 + P312 - P303 + P361 + P353 - P304 + P340 + P312 - P305 + P351 + P338