Order Entry
Northern Ireland
ContactUsLinkComponent
Organic reference standard, 2-Ethylhexyl salicylate, analytical standard, Supelco®
Organic reference standard, 2-Ethylhexyl salicylate, analytical standard, Supelco®
Catalog # 52184-1ML
Supplier:  Merck
CAS Number:  
undefined
Organic reference standard, 2-Ethylhexyl salicylate, analytical standard, Supelco®
Catalog # 52184-1ML
Supplier:  Merck
CAS Number:  

Specifications

  • Pk:
    1 mL
  • Standards:
    Analytical standard
  • CAS No.:
    118-60-5
  • Description:
    2-Ethylhexyl salicylate
  • Formula:
    C₁₅H₂₂O₃
  • Description:
    2-Ethylhexyl salicylate
  • Boiling point:
    189...190 °C/21 mmHg(lit.)

Specifications

About this item

2-Ethylhexyl salicylate may be used as a reference standard for the determination of octyl salicylate as a UV filter authorized in sunscreens by high-performance liquid chromatography.