Supelco® Organic Reference Standard, 1,4-Dioxane solution
CRM48367
: Merck
:
- Concentration:2000 µg/ml in methanol, ampule of 1 ml
- Pk:1 mL
- Standards:Certified reference material
- CAS No.:123-91-1
- Description:1,4-Dioxane solution
- Storage conditions:2...30 °C
- Hazard class description:H225 - H301 + H311 + H331 - H350 - H370
- Shipping conditions:STD
- MDL No.:MFCD00006571
- M-Clarity quality level:MQ100
- Synonym(s):1,4-Dioxane solution
- Molecular weight:88,11
- Flash point:9,7 °C - Closed cup
- GHS signal: GHS02, GHS06, GHS08
: P202 - P210 - P280 - P301 + P310 - P303 + P361 + P353 - P304 + P340 + P311