Supelco® Organic Reference Standard, 1,3-Dinonadecanoin
68633-25MG
: Merck
:
- Pk:25 mg
- Standards:Analytical standard
- CAS No.:372490-74-9
- Description:1,3-Dinonadecanoin
- Storage conditions:2...30 °C
- Shipping conditions:STD
- MDL No.:MFCD00673512
- M-Clarity quality level:MQ100
- Synonym(s):1,3-Dinonadecanoyl-rac-glycerol; 1,3-Dinonadecanoylglycerol; DG(19:0/0:0/19:0) [rac]; Glyceryl dinonadecanoate
- Molecular weight:653,07