Order Entry
Northern Ireland
ContactUsLinkComponent
 
Hydroxylammonium chloride ≥96.0% (by KMnO₄ titration), crystals, AR® ACS for the determination of mercury, Macron Fine Chemicals™
Catalog # 5258-02
CAS Number:  
undefined
Hydroxylammonium chloride ≥96.0% (by KMnO₄ titration), crystals, AR® ACS for the determination of mercury, Macron Fine Chemicals™
Catalog # 5258-02
CAS Number:  

Specifications

  • Pack type:
    PE bottle, Mall Polystormor®
  • Pk:
    125 g
  • Seq:
    3445

Specifications

Specification Test Results

  • ACS - Assay (NH₂OH·HCl) (by KMnO₄ titrn):
    ≥96.0 %
  • ACS - Clarity of Alcohol Solution:
    Passes Test
  • ACS - Residue after Ignition:
    ≤0.05 %
  • ACS - Titrable Free Acid (meq/g):
    ≤0.25
  • ACS - Ammonium (NH₄):
    Passes Test
  • ACS - Sulfur Compounds (as SO₄):
    ≤0.005 %
  • ACS - Heavy Metals (as Pb):
    ≤0.0005 %
  • ACS - Iron (Fe):
    ≤0.0005 %
  • ACS - Mercury (Hg):
    ≤0.050 ppm

Specification Test Results

About this item

 :
 :69.49 g/mol
Merck Index:13,04853
Boiling Pt:306 °C (1013 hPa)
Melting Pt:151 °C
Density:1,67 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:226-798-2
UN:2923
ADR:8 (6.1),III
 :GHS05
 :GHS06
 :GHS08
 :GHS09
CAS Number:5470-11-1
Formula:NH₂OH·HCl
MDL Number:MFCD00051089