


Leishman's eosin-methylene blue modified solution stain for histology, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:12627-53-1
:
MDL Number:MFCD00131498
Boiling Pt:65 °C (1013 hPa)
Density:0,79 g/cm3 (20 °C)
Flash Pt:12 °C
UN:1230
ADR:3 (6.1),II
:GHS02
:GHS06
:GHS08
Storage Temperature:Ambient
:1
Specification Test Results
- Suitability for microscopy (Blood smear):passes test
- Erythrocytes:reddish
- Nuclei:violet
- Eosinophilic granules:brick-red
- Neutrophilic granules:light violet
- Lymphocyte cytoplasm:Blue
Specification Test Results
Frequently Bought Together