Pesticide reference standard, Captan-d6, neat, Honeywell Fluka™
Catalog # HONCR1524-10MG
Supplier: Honeywell Chemicals
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:10 mg
- CAS No.:1330190-00-5
- Description:Captan-d₆
- Formula:C₉H₂D₆Cl₃NO₂S
- Storage temperature:Ambient
- Description:Captan-d₆
- Freezing point:174...175 °C
Specifications
About this item
High quality inorganic and organic reference materials produced according to ISO 9001.
- Ensure you have the information you need to quickly start your analysis, and pass relevant audits
- Certificate of Analysis (CoA) in every delivery complete with expiry date and lot specific analytical results