Organic analytical standards, Brilliant green hydrogen sulfate hydrate, neat, reference material, Honeywell Fluka™
HONCR1497-10MG
:
- Pack type:Glass bottle
- Pk:10 mg
- CAS No.:633-03-4
- Description:Brilliant green hydrogen sulfate hydrate
- Formula:C₂₇H₃₃N₂ · HSO₄
- Storage temperature:Ambient
- Description:Brilliant green hydrogen sulfate hydrate
- Boiling point:774 °C
- Freezing point:220 - 223 °C
High quality inorganic and organic reference materials produced according to ISO 9001.
- Ensure you have the information you need to quickly start your analysis, and pass relevant audits
- Certificate of Analysis (CoA) in every delivery complete with expiry date and lot specific analytical results