Pharmaceutical analytical standard, 1-Desoxycarbadox, reference material, Honeywell Fluka™
Catalog # HONCR1487-10MG
Supplier: Honeywell Chemicals
CAS Number:
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Specifications
- Pack type:Glass bottle
- Pk:10 mg
- CAS No.:118828-83-4
- Description:1-Desoxycarbadox
- Formula:C₁₁H₁₀N₄O₃
- Storage temperature:Ambient
- Description:1-Desoxycarbadox
- Freezing point:251...253 °C
Specifications
About this item
High quality inorganic and organic reference materials produced according to ISO 9001.
- Ensure you have the information you need to quickly start your analysis, and pass relevant audits
- Certificate of Analysis (CoA) in every delivery complete with expiry date and lot specific analytical results