Order Entry
Northern Ireland
ContactUsLinkComponent
 
Organic analytical standards, Palmitic acid glycidylester-d5, neat, reference material, Honeywell Fluka™
  HONCR1483-10MG
 :  
Organic analytical standards, Palmitic acid glycidylester-d5, neat, reference material, Honeywell Fluka™
  HONCR1483-10MG
 :  

 

  • Pack type:
    Glass bottle
  • Pk:
    10 mg
  • CAS No.:
    1794941-80-2
  • Description:
    Palmitic acid glycidylester-d₅
  • Formula:
    C₁₉H₃₁D₅O₃
  • Storage temperature:
    Ambient
  • Description:
    Palmitic acid glycidylester-d₅
  • Freezing point:
    43...44 °C

 

 

High quality inorganic and organic reference materials produced according to ISO 9001.

  • Ensure you have the information you need to quickly start your analysis, and pass relevant audits
  • Certificate of Analysis (CoA) in every delivery complete with expiry date and lot specific analytical results