Order Entry
Northern Ireland
ContactUsLinkComponent
Organic analytical standards, Dichloroisoeverninic acid-d6, neat, reference material, Honeywell Fluka™
Organic analytical standards, Dichloroisoeverninic acid-d6, neat, reference material, Honeywell Fluka™
Catalog # HONCR1480-10MG
CAS Number:  
Organic analytical standards, Dichloroisoeverninic acid-d6, neat, reference material, Honeywell Fluka™
Catalog # HONCR1480-10MG
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    10 mg
  • Description:
    Dichloroisoeverninic acid-d₆
  • Formula:
    C₉H₂D₆Cl₂O₄
  • Storage temperature:
    Ambient
  • Description:
    Dichloroisoeverninic acid-d₆
  • Freezing point:
    124...126 °C

Specifications

About this item

High quality inorganic and organic reference materials produced according to ISO 9001.

  • Ensure you have the information you need to quickly start your analysis, and pass relevant audits
  • Certificate of Analysis (CoA) in every delivery complete with expiry date and lot specific analytical results