Order Entry
Northern Ireland
ContactUsLinkComponent
 
Pharmaceutical analytical standard, Ibuprofen-d3, reference material, Honeywell Fluka™
  HONCR1470-10MG
 :  
Pharmaceutical analytical standard, Ibuprofen-d3, reference material, Honeywell Fluka™
  HONCR1470-10MG
 :  

 

  • Pack type:
    Glass bottle
  • Pk:
    10 mg
  • CAS No.:
    121662-14-4
  • Description:
    Ibuprofen-d₃
  • Formula:
    C₁₃H₁₅D₃O₂
  • Storage temperature:
    Ambient
  • Description:
    Ibuprofen-d₃
  • Freezing point:
    67...68 °C

 

 

High quality inorganic and organic reference materials produced according to ISO 9001.

  • Ensure you have the information you need to quickly start your analysis, and pass relevant audits
  • Certificate of Analysis (CoA) in every delivery complete with expiry date and lot specific analytical results