Order Entry
Northern Ireland
ContactUsLinkComponent
Pharmaceutical analytical standard, Ornidazole-13C2,15N2, reference material, Honeywell Fluka™
Pharmaceutical analytical standard, Ornidazole-13C2,15N2, reference material, Honeywell Fluka™
Catalog # HONCR1452-10MG
CAS Number:  
Pharmaceutical analytical standard, Ornidazole-13C2,15N2, reference material, Honeywell Fluka™
Catalog # HONCR1452-10MG
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    10 mg
  • Description:
    Ornidazole-¹³C₂,¹⁵N₂
  • Formula:
    C₅¹³C₂H₁₀ClN¹⁵N₂O₃
  • Storage temperature:
    Ambient
  • Description:
    Ornidazole-¹³C₂,¹⁵N₂
  • Freezing point:
    69...70 °C

Specifications

About this item

  • Ensure you have the information you need to quickly start your analysis, and pass relevant audits
  • High quality inorganic and organic reference materials produced according to ISO 9001
  • Certificate of Analysis (CoA) in every delivery complete with expiry date and lot specific analytical results