Pharmaceutical analytical standard, Ronidazole, reference material, Honeywell Fluka™
HONCR1447-50MG
:
- Pack type:Glass bottle
- Pk:50 mg
- CAS No.:7681-76-7
- Description:Ronidazole
- Formula:C₆H₈N₄O₄
- Storage temperature:Ambient
- Description:Ronidazole
- Freezing point:166...167 °C
- Ensure you have the information you need to quickly start your analysis, and pass relevant audits
- High quality inorganic and organic reference materials produced according to ISO 9001
- Certificate of Analysis (CoA) in every delivery complete with expiry date and lot specific analytical results