Pharmaceutical analytical standard, 4-NP-SCA, reference material, Honeywell Fluka™
HONCR1444-10MG
:
- Pack type:Glass bottle
- Pk:10 mg
- CAS No.:5315-87-7
- Description:4-NP-SCA
- Formula:C₈H₈N₄O₃
- Storage temperature:Ambient
- Description:4-NP-SCA
- Freezing point:205...208 °C
- Ensure you have the information you need to quickly start your analysis, and pass relevant audits
- High quality inorganic and organic reference materials produced according to ISO 9001
- Certificate of Analysis (CoA) in every delivery complete with expiry date and lot specific analytical results