Pharmaceutical analytical standard, 4-Aminoantipyrine, reference material, Honeywell Fluka™
Catalog # HONCR1425-50MG
Supplier: Honeywell Chemicals
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:50 mg
- CAS No.:83-07-8
- Description:4-Aminoantipyrine
- Formula:C₁₁H₁₃N₃O
- Storage temperature:Ambient
- Description:4-Aminoantipyrine
- Freezing point:106...111 °C
Specifications
About this item
High quality inorganic and organic reference materials produced according to ISO 9001.
- Ensure you have the information you need to quickly start your analysis, and pass relevant audits
- Certificate of Analysis (CoA) in every delivery complete with expiry date and lot specific analytical results