Pharmaceutical analytical standard, 4-Aminoantipyrine-d3, reference material, Honeywell Fluka™
HONCR1420-10MG
:
- Pack type:Glass bottle
- Pk:10 mg
- CAS No.:68229-55-0
- Description:4-Aminoantipyrine-d₃
- Formula:C₁₁H₁₀D₃N₃O
- Storage temperature:Ambient
- Description:4-Aminoantipyrine-d₃
- Freezing point:105...110 °C
High quality inorganic and organic reference materials produced according to ISO 9001.
- Ensure you have the information you need to quickly start your analysis, and pass relevant audits
- Certificate of Analysis (CoA) in every delivery complete with expiry date and lot specific analytical results