Pharmaceutical analytical standard, Phenylethanolamine A-d3, reference material, Honeywell Fluka™
Catalog # HONCR1413-10MG
Supplier: Honeywell Chemicals
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:10 mg
- Description:Phenylethanolamine A-d₃
- Formula:C₁₉H₂₁D₃N₂O₄
- Storage temperature:Ambient
- Description:Phenylethanolamine A-d₃
- Freezing point:80...81 °C
Specifications
About this item
High quality inorganic and organic reference materials produced according to ISO 9001.
- Ensure you have the information you need to quickly start your analysis, and pass relevant audits
- Certificate of Analysis (CoA) in every delivery complete with expiry date and lot specific analytical results