Organic analytical standards, Sulfisozole sodium, neat, reference material, Honeywell Fluka™
Catalog # HONCR1253-25MG
Supplier: Honeywell Chemicals
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:25 mg
- CAS No.:37514-39-9
- Description:Sulfisozole sodium
- Formula:C₉H₈N₃NaO₃S
- Storage temperature:Ambient
- Description:Sulfisozole sodium
- Freezing point:276...279 °C
Specifications
About this item
High quality inorganic and organic reference materials produced according to ISO 9001.
- Ensure you have the information you need to quickly start your analysis, and pass relevant audits
- Certificate of Analysis (CoA) in every delivery complete with expiry date and lot specific analytical results