Pharmaceutical analytical standard, Nifurpirinol, reference material, Honeywell Fluka™
Catalog # HONCR1234-50MG
Supplier: Honeywell Chemicals
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:50 mg
- CAS No.:13411-16-0
- Description:Nifurpirinol
- Formula:C₁₂H₁₀N₂O₄
- Storage temperature:Ambient
- Description:Nifurpirinol
- Freezing point:170...171 °C
Specifications
About this item
High quality inorganic and organic reference materials produced according to ISO 9001.
- Ensure you have the information you need to quickly start your analysis, and pass relevant audits
- Certificate of Analysis (CoA) in every delivery complete with expiry date and lot specific analytical results