Pharmaceutical analytical standard, Luxabendazole-(methyl-d3), reference material, Honeywell Fluka™
HONCR1152-10MG
:
- Pack type:Glass bottle
- Pk:10 mg
- CAS No.:1448346-27-7
- Description:Luxabendazole-(methyl-d₃)
- Formula:C₁₅D₃H₉FN₃O₅S
- Storage temperature:Ambient
- Description:Luxabendazole-(methyl-d₃)
- Freezing point:221...223 °C
High quality inorganic and organic reference materials produced according to ISO 9001.
- Ensure you have the information you need to quickly start your analysis, and pass relevant audits
- Certificate of Analysis (CoA) in every delivery complete with expiry date and lot specific analytical results